FLUORESCENT DYE 350-I PHALLOIDIN FLUORESCENCE DYE 350-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 300 Reactions 1 * 300 Essai
Fournisseur: Abnova
FLUORESCENT DYE 350-I PHALLOIDIN FLUORESCENCE DYE 350-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 300 Reactions 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
FLUORESCENT DYE 350-I PHALLOIDIN FLUORESCENCE DYE 350-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 300 Reactions 1 * 300 Essai ABNOU0276 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou