Fluorescence Dye 555-I with Phalloidin conjugate. Compared with Cy3 conjugates, those of Fluorescence Dye 555-I are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Beside 1 * 300 Essai
Fournisseur: Abnova
Fluorescence Dye 555-I with Phalloidin conjugate. Compared with Cy3 conjugates, those of Fluorescence Dye 555-I are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Beside 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
Fluorescence Dye 555-I with Phalloidin conjugate. Compared with Cy3 conjugates, those of Fluorescence Dye 555-I are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Beside 1 * 300 Essai ABNOU0289 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou