FLUORESCENT DYE 594-I PHALLOIDIN FLUORESCENCE DYE 594-I WITH PHALLOIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594- I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE OF 1 * 300 Essai
Fournisseur: Abnova
FLUORESCENT DYE 594-I PHALLOIDIN FLUORESCENCE DYE 594-I WITH PHALLOIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594- I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE OF 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
FLUORESCENT DYE 594-I PHALLOIDIN FLUORESCENCE DYE 594-I WITH PHALLOIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594- I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE OF 1 * 300 Essai ABNOU0292 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou