FLUORESCENT DYE 647-I PHALLOIDIN FLUORESCENCE DYE 647-I WITH PHALLOIDIN CONJUGATE. THE SLIGHT RED SHIFT IN ABSORPTION SPECTRUM MAKE S FLUORESCENCE DYE 647-I AN OPTIMAL MATCH TO FILTERS DESIGNED CY5 DYES.BESIDES, THEIR FLUORESCENCE IS PH-INDEPENDENT 1 * 300 Essai
Fournisseur: Abnova
FLUORESCENT DYE 647-I PHALLOIDIN FLUORESCENCE DYE 647-I WITH PHALLOIDIN CONJUGATE. THE SLIGHT RED SHIFT IN ABSORPTION SPECTRUM MAKE S FLUORESCENCE DYE 647-I AN OPTIMAL MATCH TO FILTERS DESIGNED CY5 DYES.BESIDES, THEIR FLUORESCENCE IS PH-INDEPENDENT 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
FLUORESCENT DYE 647-I PHALLOIDIN FLUORESCENCE DYE 647-I WITH PHALLOIDIN CONJUGATE. THE SLIGHT RED SHIFT IN ABSORPTION SPECTRUM MAKE S FLUORESCENCE DYE 647-I AN OPTIMAL MATCH TO FILTERS DESIGNED CY5 DYES.BESIDES, THEIR FLUORESCENCE IS PH-INDEPENDENT 1 * 300 Essai ABNOU0298 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou