FLUORESCENT DYE 700-I PHALLOIDIN FLUORESCENCE DYE 700-I WITH PHALLOIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESCENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WELL SE 1 * 300 Essai
Fournisseur: Abnova
FLUORESCENT DYE 700-I PHALLOIDIN FLUORESCENCE DYE 700-I WITH PHALLOIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESCENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WELL SE 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
FLUORESCENT DYE 700-I PHALLOIDIN FLUORESCENCE DYE 700-I WITH PHALLOIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESCENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WELL SE 1 * 300 Essai ABNOU0302 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou