FLUORESCENT DYE 750-I PHALLOIDIN FLUORESCENCE DYE 750-I WITH PHALLOIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT DYE 7 50-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED FRO 1 * 300 Essai
Fournisseur: Abnova
FLUORESCENT DYE 750-I PHALLOIDIN FLUORESCENCE DYE 750-I WITH PHALLOIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT DYE 7 50-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED FRO 1 * 300 Essai
COMMANDE
Développer tout / Réduire tout
FLUORESCENT DYE 750-I PHALLOIDIN FLUORESCENCE DYE 750-I WITH PHALLOIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT DYE 7 50-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED FRO 1 * 300 Essai ABNOU0305 1 * 300 Essai
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou