Avantor® ACE® PFAS Delay Columns
Fournisseur: Avantor
ACE-PFASD-0502EA 698 EUR InStock
ACE-PFASD-0502
Avantor® ACE® PFAS Delay Columns
Colonnes pour chromatographie
Background PFAS (Perfluoronated Alkyl Substances) from an LC instrument and mobile phase can interfere with the detection of PFAS in samples. The introduction of a delay column, which is plumbed before the autosampler in the LC system, will delay background PFAS to improve accuracy of PFAS measurement by LC-MS.​
  • Silica-based fully porous particle delay column
  • Universal, can be used in a workflow setup with any analytical column format/dimensions
Allows for the analysis of PFAS from the sample, without interfering PFAS compromising the analysis (present and introduced by the instrumentation). Moreover, the data output/chromatogram benefits with the respective PFAS sample peaks separated/delayed from the background PFAS peaks.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité