Human 2B4, His Tag (2B4-H5224) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human 2B4, His Tag (2B4-H5224) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human 2B4, His Tag (2B4-H5224) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 1 mg ACRB2B4-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou