Cynomolgus Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (A ccession XP005544945.1). In the region Leu 24 - Gln 186, the AA sequence of Cynomolgus and Rhesus macaque 4-1BB are h 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (A ccession XP005544945.1). In the region Leu 24 - Gln 186, the AA sequence of Cynomolgus and Rhesus macaque 4-1BB are h 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque 4-1BB, His Tag (41B-C52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (A ccession XP005544945.1). In the region Leu 24 - Gln 186, the AA sequence of Cynomolgus and Rhesus macaque 4-1BB are h 1 * 100 µG ACRB41B-C52H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou