Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human Latent Activin A, His Tag (ACA-H424x) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ser 426 (Accession AAH07858.1). 1 * 100 µG ACRBACA-H424X100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou