Recombinat Protein,Mouse ADAM17 TACE CD156b Protein, His Tag,Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Asp 563 (Accession Q9Z0F8).This protein carries a polyhistidine tag at the C-terminus 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse ADAM17 TACE CD156b Protein, His Tag,Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Asp 563 (Accession Q9Z0F8).This protein carries a polyhistidine tag at the C-terminus 1 * 50 µG ACRBAD7-M52H1-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou