Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession AAI15405.1 ). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession AAI15405.1 ). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession AAI15405.1 ). 1 * 50 µG ACRBAD8-H5223-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou