Recombinat Protein,Human Akt1 Protein, His,Strep II Tag,Human Akt1, His,Strep II Tag (AK1-H5283) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Ala 480 (Accession AAH00479).This protein carries a polyhistidine tag at the C-termi 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Akt1 Protein, His,Strep II Tag,Human Akt1, His,Strep II Tag (AK1-H5283) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Ala 480 (Accession AAH00479).This protein carries a polyhistidine tag at the C-termi 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Akt1 Protein, His,Strep II Tag,Human Akt1, His,Strep II Tag (AK1-H5283) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Ala 480 (Accession AAH00479).This protein carries a polyhistidine tag at the C-termi 1 * 500 µG ACRBAK1-H5283500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou