Recombinat Protein,Biotinylated Human Angiopoietin-2 ANGPT2 Protein, His,Avitag,Biotinylated Human Angiopoietin-2, His,Avitag (AN2 -H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This prot 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human Angiopoietin-2 ANGPT2 Protein, His,Avitag,Biotinylated Human Angiopoietin-2, His,Avitag (AN2 -H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This prot 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human Angiopoietin-2 ANGPT2 Protein, His,Avitag™,Biotinylated Human Angiopoietin-2, His,Avitag (AN2 -H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession AAI26201.1).This prot 1 * 25 µG ACRBAN2-H82E7-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou