Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Angiopoietin-like 3 ANGPTL3 Protein, His Tag,Human Angiopoietin-like 3, His Tag (AN3-H5226) is expressed f rom human 293 cells (HEK293). It contains AA Ser 17 - Pro 220 (Accession NP055310).This protein carries a polyhistidi 1 * 1 mg ACRBAN3-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou