Recombinat Protein,Human Angiopoietin-like 4 ANGPTL4 (166-406) Protein, His Tag,Human Angiopoietin-like 4 (166-406), His Tag (AN4-H 5243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession NP647475).This protein h 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 4 ANGPTL4 (166-406) Protein, His Tag,Human Angiopoietin-like 4 (166-406), His Tag (AN4-H 5243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession NP647475).This protein h 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Angiopoietin-like 4 ANGPTL4 (166-406) Protein, His Tag,Human Angiopoietin-like 4 (166-406), His Tag (AN4-H 5243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession NP647475).This protein h 1 * 100 µG ACRBAN4-H5243100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou