Recombinat Protein,Human Annexin A5 Protein, His Tag,Human Annexin A5, His Tag (AN5-H5123) is expressed from E.coli cells. It contai ns AA Met 1 - Asp 320 (Accession P08758-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Annexin A5 Protein, His Tag,Human Annexin A5, His Tag (AN5-H5123) is expressed from E.coli cells. It contai ns AA Met 1 - Asp 320 (Accession P08758-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG ACRBAN5-H5123100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou