Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Apo-A1 Protein, His Tag,Human Apo-A1, His Tag (AP1-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Arg 19 - Gln 267 (Accession NP000030.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg ACRBAP1-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou