Recombinat Protein,Human ApolipoproteinA-II APOA2 Protein, His Tag,Human ApolipoproteinA-II, His Tag (APA-H51H6) is expressed fro m E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the e 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human ApolipoproteinA-II APOA2 Protein, His Tag,Human ApolipoproteinA-II, His Tag (APA-H51H6) is expressed fro m E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the e 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Apolipoprotein A-II APOA2 Protein, His Tag,Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed fro m E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the e 1 * 1 mg ACRBAPA-H51H6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou