Recombinat Protein,Human Arginase 1 ARG1 Protein, His Tag,Human Arginase 1, His Tag (AR1-H5228) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Lys 322 (Accession NP000036).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Arginase 1 ARG1 Protein, His Tag,Human Arginase 1, His Tag (AR1-H5228) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Lys 322 (Accession NP000036).This protein carries a polyhistidine tag at the C-termin 1 * 1 mg ACRBAR1-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou