Recombinat Protein,Cynomolgus Axl Protein, His Tag,Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). I t contains AA Pro 27 - Pro 293 (Accession XP005589384).This protein carries a polyhistidine tag at the C-terminus. T 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus Axl Protein, His Tag,Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). I t contains AA Pro 27 - Pro 293 (Accession XP005589384).This protein carries a polyhistidine tag at the C-terminus. T 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus Axl Protein, His Tag,Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). I t contains AA Pro 27 - Pro 293 (Accession XP005589384).This protein carries a polyhistidine tag at the C-terminus. T 1 * 100 µG ACRBAXL-C52H3100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou