Recombinat Protein,Human Axl Protein, His Tag,Human Axl, His Tag (AXL-H5226) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 449 (Accession AAH32229).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Axl Protein, His Tag,Human Axl, His Tag (AXL-H5226) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 449 (Accession AAH32229).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Axl Protein, His Tag,Human Axl, His Tag (AXL-H5226) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 449 (Accession AAH32229).This protein carries a polyhistidine tag at the C-terminus. The protein ha M 1 * 1 mg ACRBAXL-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou