Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession P25213), was produced in human 293 cells (HEK293) at ACROBiosystems. 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession P25213), was produced in human 293 cells (HEK293) at ACROBiosystems. 1 * 50 µG ACRBB1R-H52H6-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou