Recombinat Protein,Human beta 2-Microglobulin B2M Protein, His Tag,Human B2M, His Tag (B2M-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession AAH32589).This protein carries a polyhistidine tag at the C-ter t 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human beta 2-Microglobulin B2M Protein, His Tag,Human B2M, His Tag (B2M-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession AAH32589).This protein carries a polyhistidine tag at the C-ter t 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human beta 2-Microglobulin B2M Protein, His Tag,Human B2M, His Tag (B2M-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (Accession AAH32589).This protein carries a polyhistidine tag at the C-ter t 1 * 1 mg ACRBB2M-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou