Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP0055 48617.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP0055 48617.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP0055 48617.1). 1 * 1 mg ACRBB72-C52HB-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou