Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession XP0153 08534.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession XP0153 08534.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus B7-H3, His Tag (B73-C52Ha) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Glu 465 (Accession XP0153 08534.1). 1 * 100 µG ACRBB73-C52HA100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou