Recombinat Protein,Biotinylated Human B7-H5 Gi24 VISTA Protein, Fc,Avitag,Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expr essed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568.1).This protein carries a hum 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human B7-H5 Gi24 VISTA Protein, Fc,Avitag,Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expr essed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568.1).This protein carries a hum 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human B7-H5 Gi24 VISTA Protein, Fc,Avitag™,Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expr essed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568.1).This protein carries a hum 1 * 200 µG ACRBB75-H82F3200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou