Recombinat Protein,Human BACE-1 Protein, Tag Free,Human BACE-1, Tag Free (BA1-H5213) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries no tag. The protein has a calculated MW of 4 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human BACE-1 Protein, Tag Free,Human BACE-1, Tag Free (BA1-H5213) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries no tag. The protein has a calculated MW of 4 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human BACE-1 Protein, Tag Free,Human BACE-1, Tag Free (BA1-H5213) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries no tag. The protein has a calculated MW of 4 1 * 100 µG ACRBBA1-H5213100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou