Recombinat Protein,Human BACE-1 Protein, His Tag,Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It co ntains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human BACE-1 Protein, His Tag,Human BACE-1, His Tag (BA1-H5220) is expressed from human 293 cells (HEK293). It co ntains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg ACRBBA1-H5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou