Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human BAFF, His,Avitag (active trimer) (MALS verified) (BAF-H82Q2) is expressed from human 293 cells (HEK293). It conta ins AA Ala 134 - Leu 285 (Accession AAH20674.1). 1 * 25 µG ACRBBAF-H82Q2-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou