Recombinat Protein,Human Bcl-x BCL2L1 Protein, His Tag,Human Bcl-x, His Tag (BC1-H5120) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession AAH19307).This protein carries a polyhistidine tag at the C-terminus. The protein has W 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Bcl-x BCL2L1 Protein, His Tag,Human Bcl-x, His Tag (BC1-H5120) is expressed from E.coli cells. It contains AA Met 1 - Arg 212 (Accession AAH19307).This protein carries a polyhistidine tag at the C-terminus. The protein has W 1 * 100 µG ACRBBC1-H5120100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou