Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA a 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA a 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Cynomolgus Rhesus macaque BCMA, Fc,Avitag (BCA-C82F4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA a 1 * 25 µG ACRBBCA-C82F4-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou