Recombinat Protein,Human Betacellulin Protein, Fc Tag,Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK2 93). It contains AA Asp 32 - Tyr 111 (Accession AAH11618).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Betacellulin Protein, Fc Tag,Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK2 93). It contains AA Asp 32 - Tyr 111 (Accession AAH11618).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Betacellulin Protein, Fc Tag,Human Betacellulin, Fc Tag (BEN-H525x) is expressed from human 293 cells (HEK2 93). It contains AA Asp 32 - Tyr 111 (Accession AAH11618).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg ACRBBEN-H525X-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou