Recombinat Protein,Biotinylated Human BTN1A1 Butyrophilin Protein, His,Avitag,Biotinylated Human BTN1A1, His,Avitag (BT1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession Q13410-1).This protein carries a 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human BTN1A1 Butyrophilin Protein, His,Avitag,Biotinylated Human BTN1A1, His,Avitag (BT1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession Q13410-1).This protein carries a 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human BTN1A1 Butyrophilin Protein, His,Avitag™,Biotinylated Human BTN1A1, His,Avitag (BT1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession Q13410-1).This protein carries a 1 * 25 µG ACRBBT1-H82E6-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou