Recombinat Protein,Biotinylated Human BTN3A1 CD277 Protein, Fc,Avitag,Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expresse d from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession O00481-1).This protein carries a human Ig 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human BTN3A1 CD277 Protein, Fc,Avitag,Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expresse d from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession O00481-1).This protein carries a human Ig 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human BTN3A1 CD277 Protein, Fc,Avitag™,Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expresse d from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession O00481-1).This protein carries a human Ig 1 * 200 µG ACRBBT1-H82F7200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou