Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 100 µG ACRBBT3-H52H0100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou