Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 µG ACRBBTA-M5253100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou