Human CD160, His Tag (BY5-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 159 (Accession O95971-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CD160, His Tag (BY5-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 159 (Accession O95971-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CD160, His Tag (BY5-H5229) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 159 (Accession O95971-1). 1 * 1 mg ACRBBY5-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou