Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 µG ACRBC5A-M51H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou