Recombinat Protein,Human Carbonic Anhydrase II CA2 Protein, His Tag,Human CA2, His Tag (CA2-H5228) is expressed from human 293 cell s (HEK293). It contains AA Met 1 - Lys 260 (Accession NP000058.1).This protein carries a polyhistidine tag at the C-t 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Carbonic Anhydrase II CA2 Protein, His Tag,Human CA2, His Tag (CA2-H5228) is expressed from human 293 cell s (HEK293). It contains AA Met 1 - Lys 260 (Accession NP000058.1).This protein carries a polyhistidine tag at the C-t 1 * 100 µG ACRBCA2-H5228100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou