Recombinat Protein,Human Cadherin-6 KCAD CDH6 Protein, His Tag,Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 c ells (HEK293). It contains AA Thr 19 - Ala 615 (Accession AAH00019).This protein carries a polyhistidine tag at the C- 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Cadherin-6 KCAD CDH6 Protein, His Tag,Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 c ells (HEK293). It contains AA Thr 19 - Ala 615 (Accession AAH00019).This protein carries a polyhistidine tag at the C- 1 * 1 mg ACRBCA6-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou