Recombinat Protein,Human Carbonic Anhydrase IX CA9 (138-414) Protein, His Tag,Human Carbonic Anhydrase IX (138-414), His Tag (CA9-H 5220) is expressed from human 293 cells (HEK293). It contains AA Gln 138 - Asp 414 (Accession NP001207.2).This protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Carbonic Anhydrase IX CA9 (138-414) Protein, His Tag,Human Carbonic Anhydrase IX (138-414), His Tag (CA9-H 5220) is expressed from human 293 cells (HEK293). It contains AA Gln 138 - Asp 414 (Accession NP001207.2).This protei 1 * 1 mg ACRBCA9-H5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou