Cynomolgus Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Acc ession G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Acc ession G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque B7-1, Fc Tag (CD0-C5252) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Acc ession G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. 1 * 100 µG ACRBCD0-C5252100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou