Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38) . 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38) . 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38) . 1 * 1 mg ACRBCD0-C52H6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou