Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 20 G
Fournisseur: ACROBIOSYSTEMS
Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 20 G
COMMANDE
Développer tout / Réduire tout
Human CD20 Full Length, His Tag (HEK293) (SPR verified) (CD0-H52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession P11836-1). 1 * 20 µG ACRBCD0-H52H3-20UG 1 * 20 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou