Recombinat Protein,Human CD31 PECAM-1 Protein, His Tag,Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession P16284-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD31 PECAM-1 Protein, His Tag,Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession P16284-1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg ACRBCD1-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou