Recombinat Protein,Human CD31 PECAM-1 Protein, Fc Tag,Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession AAH22512.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD31 PECAM-1 Protein, Fc Tag,Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession AAH22512.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD31 PECAM-1 Protein, Fc Tag,Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession AAH22512.1).This protein carries a human IgG1 Fc tag at the C-terminus. The ca 1 * 200 µG ACRBCD1-H5258200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou