Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (A ccession NP000869.1). 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (A ccession NP000869.1). 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (A ccession NP000869.1). 1 * 500 µG ACRBCD2-H5221500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou