Recombinat Protein,Human CD83 Protein, His Tag,Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Ala 143 (Accession AAH30830).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD83 Protein, His Tag,Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Ala 143 (Accession AAH30830).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD83 Protein, His Tag,Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Ala 143 (Accession AAH30830).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBCD3-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou