Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152 .1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152 .1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152 .1). 1 * 1 mg ACRBCD3-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou